5-Chlorosalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: CSA, 5-Chloro-2-hydroxybenzoic acid

C0287-500G C0287-25G
TCC0287-500GEA 122 USD
TCC0287-500G TCC0287-25G
5-Chlorosalicylic acid ≥98.0% (by GC, titration analysis)
5-Chlorosalicylic acid

CAS Number: 321-14-2
MDL Number: MFCD00002457
Molecular Formula: C7H5ClO3
Molecular Weight: 172.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 5-Chloro-2-hydroxybenzoic Acid

Formula: C₇H₅ClO₃
MW: 172.57 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00002457
CAS Number: 321-14-2
EINECS: 206-283-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfbc394048367ab638101007c2f6fdba3672153c&vid=76bcd473af4489c1ab29989ec1b9945463ee6777&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR