3-Chloropropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Chloropropanoic acid
C0276-500G
C0276-25G
TCC0276-500GEA
87.48
USD
TCC0276-500G
TCC0276-25G
3-Chloropropionic acid ≥98.0% (by GC, titration analysis)
3-Chloropropionic acid
CAS Number: 107-94-8
MDL Number: MFCD00002764
Molecular Formula: C3H5ClO2
Molecular Weight: 108.52
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 40
Flash Point (°C): 110
Storage Temperature: 0-10°C
Formula:
C₃H₅ClO₂ MW: 108.52 g/mol Boiling Pt: 200 °C Melting Pt: 40 °C Density: 1.26 g/cm³ (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002764 CAS Number: 107-94-8 EINECS: 203-534-4 UN: 3261 ADR: 8,II Merck Index: 13,02178 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...