3-Chloro-1-propanol ≥98.0%

Supplier: TCI America

Synonyms: 3-Chloropropan-1-ol, 1-Chloro-3-hydroxypropane

C0270-25G C0270-100G
TCC0270-25GEA 39.62 USD
TCC0270-25G TCC0270-100G
3-Chloro-1-propanol ≥98.0%
3-Chloro-1-propanol

CAS Number: 627-30-5
MDL Number: MFCD00002943
Molecular Formula: C3H7ClO
Molecular Weight: 94.54
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 82
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: Trimethylene Chlorohydrin

Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 160 °C
Melting Pt: -20 °C
Density: 1.14 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002943
CAS Number: 627-30-5
EINECS: 210-992-9
UN: 2849
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e3bc28bbde2ae9009c517902ab73cf07ac4d57c&vid=cc18e094de5aa98eddc8f67a9776bb5dbe9b204b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR