Monuron (N-(4-chlorophenyl)-N',N'-dimethylurea) ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

C0254-500G C0254-25G
TCC0254-500GEA 273 USD
TCC0254-500G TCC0254-25G
Monuron (N-(4-chlorophenyl)-N',N'-dimethylurea) ≥95.0% (by HPLC, total nitrogen)
Monuron (N-(4-chlorophenyl)-N',N'-dimethylurea)

CAS Number: 150-68-5
MDL Number: MFCD00018556
Molecular Formula: C9H11ClN2O
Molecular Weight: 198.65
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal
Melting point (°C): 173
Lambda max.: 244 nm (H2O)


Other Product Names / Synonyms: 1,1-Dimethyl-3-(4-chlorophenyl)urea, Monuron

Formula: C₉H₁₁ClN₂O
MW: 198.65
Melting Pt: 173 °C
MDL Number: MFCD00018556
CAS Number: 150-68-5
EINECS: 205-766-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=974bfc7f1451e5606f158dacfd885c7ce90b3c53&vid=974bfc7f1451e5606f158dacfd885c7ce90b3c53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR