4-Chlorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: p-Chlorophenol

C0243-500G C0243-25G
TCC0243-500GEA 53.13 USD
TCC0243-500G TCC0243-25G
4-Chlorophenol ≥98.0% (by GC, titration analysis)
4-Chlorophenol

CAS Number: 106-48-9
MDL Number: MFCD00002318
Molecular Formula: C6H5ClO
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 220
Flash Point (°C): 121
Freezing point (°C): 43

Formula: C₆H₅ClO
MW: 128.56 g/mol
Boiling Pt: 220 °C
Melting Pt: 41…44 °C
Density: 1.31 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00002318
CAS Number: 106-48-9
EINECS: 203-402-6
UN: 2020
ADR: 6.1,III
Merck Index: 13,02173
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdb9bdb02c6f516ae80f260df108812bf6b4c618&vid=754a177b52fecdf7254f94608714b74142533e05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR