3-Chlorophenol ≥98.0%
Supplier: TCI America
|
Warning
|
C0242-500G
C0242-25G
C0242-100G
TCC0242-500GEA
307
USD
TCC0242-500G
TCC0242-25G
TCC0242-100G
3-Chlorophenol ≥98.0%
3-Chlorophenol
CAS Number: 108-43-0
MDL Number: MFCD00002256
Molecular Formula: C6H5ClO
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 214
Flash Point (°C): 112
Freezing point (°C): 33
Formula:
C₆H₅ClO Boiling Pt: 214 °C Melting Pt: 31…34 °C Density: 1.245 g/cm³ (20 °C) Flash Pt: 112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002256 CAS Number: 108-43-0 EINECS: 203-582-6 UN: 2020 ADR: 6.1,III Merck Index: 13,02173 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...