1-Chlorooctane ≥99.0%
Supplier: TCI America
|
Warning
|
CAS Number: 111-85-3
MDL Number: MFCD00001022
Molecular Formula: C8H17Cl
Molecular Weight: 148.67
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 64
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: n-Octyl Chloride
Formula:
C₈H₁₇Cl Boiling Pt: 183 °C Melting Pt: –58 °C Density: 0.88 (20 °C) Flash Pt: 64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001022 CAS Number: 111-85-3 EINECS: 203-915-5 UN: 3082 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...