4-Chloro-3-nitrobenzotrifluoride ≥97.0%

Supplier: TCI America
Warning

Synonyms: 1-Chloro-2-nitro-4-(trifluoromethyl)benzene, 4-Chloro-α-α-α-trifluoro-3-nitrotoluene, 4-Chloro-3-nitrobenzotrifluoride

C0225-500G C0225-25G
TCC0225-500GEA 94 USD
TCC0225-500G TCC0225-25G
4-Chloro-3-nitrobenzotrifluoride ≥97.0%
4-Chloro-3-nitrobenzotrifluoride

CAS Number: 121-17-5
MDL Number: MFCD00007084
Molecular Formula: C7H3ClF3NO2
Molecular Weight: 225.55
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 101
Specific Gravity (20/20): 1.54


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Chloro-2-nitro-4-(trifluoromethyl)benzene

Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 222 °C
Melting Pt: -3 °C
Density: 1.54 (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007084
CAS Number: 121-17-5
EINECS: 204-451-6
UN: 2307
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19ccff8f08c9afd681505eb46dd0eb90a10e611f&vid=250989aaf4530e506b59db3c679f8d817e323473&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR