1-Chloro-4-nitrobenzene ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
4-Chloronitrobenzene
C0222-500G
C0222-25G
TCC0222-500GEA
30.52
USD
TCC0222-500G
TCC0222-25G
1-Chloro-4-nitrobenzene ≥98.0%
1-Chloro-4-nitrobenzene
CAS Number: 100-00-5
MDL Number: MFCD00007285
Molecular Formula: C6H4ClNO2
Molecular Weight: 157.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 242
Melting point (°C): 84
Flash Point (°C): 127
Other Product Names / Synonyms: 1-Chloro-4-nitrobenzene
Formula:
C₆H₄ClNO₂ MW: 157.56 g/mol Boiling Pt: 242 °C Melting Pt: 84 °C Density: 1.52 g/cm³ (20 °C) Flash Pt: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007285 CAS Number: 100-00-5 EINECS: 202-809-6 UN: 1578 ADR: 6.1,II Merck Index: 13,02170 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...