3-Chloro-2-methylpropene ≥98.0%

Supplier: TCI America
Danger

C0209-500ML C0209-25ML
TCC0209-500MLEA 40.67 USD
TCC0209-500ML TCC0209-25ML
3-Chloro-2-methylpropene ≥98.0%
3-Chloro-2-methylpropene

CAS Number: 563-47-3
MDL Number: MFCD00000953
Molecular Formula: C4H7Cl
Molecular Weight: 90.55
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 73
Flash Point (°C): -12
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: 3-Chloroisobutene, beta-Methallyl Chloride, 2-Methylallyl Chloride, 2-Methyl-2-propenyl Chloride

Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 73 °C
Melting Pt: -80 °C
Density: 0.93 (20 °C)
Flash Pt: -12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000953
CAS Number: 563-47-3
EINECS: 209-251-2
UN: 2554
ADR: 3,II
Merck Index: 13,02167
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a060aa83c37f19a9d81410c6b149d8a321411872&vid=9a1b69422e23dd7495b8747ab81c7e12865d4a78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR