2-Chloro-2-methylpropane ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
tert-Butyl chloride
C0208-500ML
C0208-25ML
TCC0208-500MLEA
75.93
USD
TCC0208-500ML
TCC0208-25ML
2-Chloro-2-methylpropane ≥98.0%
2-Chloro-2-methylpropane
CAS Number: 507-20-0
MDL Number: MFCD00000816
Molecular Formula: C4H9Cl
Molecular Weight: 92.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 52
Flash Point (°C): -18
Specific Gravity (20/20): 0.84
Other Product Names / Synonyms: tert-Butyl Chloride
Formula:
C₄H₉Cl MW: 92.57 g/mol Boiling Pt: 52 °C Melting Pt: –28 °C Density: 0.84 (20 °C) Flash Pt: –18 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000816 CAS Number: 507-20-0 EINECS: 208-066-4 UN: 1127 ADR: 3,II Merck Index: 13,01561 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...