1-Chloro-2-methylpropane ≥98.0%

Supplier: TCI America

C0207-500ML C0207-25ML
TCC0207-500MLEA 224.96 USD
TCC0207-500ML TCC0207-25ML
1-Chloro-2-methylpropane ≥98.0%
1-Chloro-2-methylpropane

CAS Number: 513-36-0
MDL Number: MFCD00000954
Molecular Formula: C4H9Cl
Molecular Weight: 92.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): -6
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Isobutyl Chloride

Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 70 °C
Melting Pt: -131 °C
Density: 0.88 (20 °C)
Flash Pt: -6 °C
Storage Temperature: Ambient
MDL Number: MFCD00000954
CAS Number: 513-36-0
EINECS: 208-157-9
UN: 1127
ADR: 3,II
Merck Index: 13,05155
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1aa2ca8d1d572a521e5defbed9fb32093aaba04&vid=802a4f71aa6062c0b509660a2e5fa6bcb3e248e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR