1-Chloro-3-methylbutane ≥98.0%

Supplier: TCI America

Synonyms: 3-Methylbutylchloride

C0199-500ML C0199-25ML
TCC0199-500MLEA 601 USD
TCC0199-500ML TCC0199-25ML
1-Chloro-3-methylbutane ≥98.0%
1-Chloro-3-methylbutane

CAS Number: 107-84-6
MDL Number: MFCD00013693
Molecular Formula: C5H11Cl
Molecular Weight: 106.59
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 16
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Isoamyl Chloride

Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 100 °C
Melting Pt: –104.4 °C
Density: 0.88 (20 °C)
Flash Pt: 16 °C
MDL Number: MFCD00013693
CAS Number: 107-84-6
EINECS: 203-525-5
UN: 1107
ADR: 3,II
Merck Index: 13,05132
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b52729e9963a7aee4fe67e1fac87807abb82e91&vid=d4ed7be4b00a62e0e96524c2cfdedf14cf282549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR