3-Chlorobutanone ≥95.0%
Supplier: TCI America
|
Danger
|
CAS Number: 4091-39-8
MDL Number: MFCD00000865
Molecular Formula: C4H7ClO
Molecular Weight: 106.55
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 23
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1-Acetyl-1-chloroethane, 1-Chloroethyl Methyl Ketone
Formula:
C₄H₇ClO Boiling Pt: 116 °C Density: 1.06 (20 °C) Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000865 CAS Number: 4091-39-8 EINECS: 223-834-9 UN: 1224 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...