4-Chloro-o-cresol ≥90.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1-Chloro-4-hydroxy-3-methylbenzene, 4-Chloro-2-methylphenol
C0152-500G
C0152-25G
TCC0152-500GEA
192
USD
TCC0152-500G
TCC0152-25G
4-Chloro-o-cresol ≥90.0%
4-Chloro-o-cresol
CAS Number: 1570-64-5
MDL Number: MFCD00002321
Molecular Formula: C7H7ClO
Molecular Weight: 142.58
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 40
Other Product Names / Synonyms: 4-Chloro-2-methylphenol
Formula:
C₇H₇ClO Boiling Pt: 224 °C (1013 hPa) Melting Pt: 46…47 °C Flash Pt: 110 °C |
MDL Number:
MFCD00002321 CAS Number: 1570-64-5 EINECS: 216-381-3 UN: 3437 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...