4-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

C0141-500G C0141-25G C0141-100G
TCC0141-500GEA 80 USD
TCC0141-500G TCC0141-25G TCC0141-100G
4-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Chlorobenzoyl chloride

CAS Number: 122-01-0
MDL Number: MFCD00000686
Molecular Formula: C7H4Cl2O
Molecular Weight: 175.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 118
Freezing point (°C): 12
Specific Gravity (20/20): 1.37

Formula: C₇H₄Cl₂O
Boiling Pt: 222 °C
Melting Pt: 14…15 °C
Density: 1.37 (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00000686
CAS Number: 122-01-0
EINECS: 204-515-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5bdce95f35dfe4f54920f0b7a6c087c5191b7ee&vid=a5bdce95f35dfe4f54920f0b7a6c087c5191b7ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR