4-Chlorothiophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol

C0129-500G C0129-25G
TCC0129-500GEA 226 USD
TCC0129-500G TCC0129-25G
4-Chlorothiophenol ≥98.0% (by GC, titration analysis)
4-Chlorothiophenol

CAS Number: 106-54-7
MDL Number: MFCD00004847
Molecular Formula: C6H5ClS
Molecular Weight: 144.62
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 206
Melting point (°C): 53
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Chloromercaptobenzene, 4-Chlorothiophenol

Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 206 °C
Melting Pt: 53 °C
Density: 1.224 g/cm³ (60 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004847
CAS Number: 106-54-7
EINECS: 203-408-9
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04f1607e57d9a876c0562147af9bd40f8c5e598f&vid=30f02ff4fcf582107970594bbf688a914721d568&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR