4-Chloroaniline ≥99.0%

Supplier: TCI America
Danger

C0112-500G C0112-25G
TCC0112-500GEA 55.95 USD
TCC0112-500G TCC0112-25G
4-Chloroaniline ≥99.0%
4-Chloroaniline

CAS Number: 106-47-8
MDL Number: MFCD00007835
Molecular Formula: C6H6ClN
Molecular Weight: 127.57
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 232
Melting point (°C): 72
Flash Point (°C): 105

Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 232 °C
Melting Pt: 72 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 13,02136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64db6904619cb3b029dc20bc7c4747c26a9e0305&vid=b91f0ed330fd3dee583aeccf7034eaea716fb570&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR