3-Chloroaniline ≥99.0%

Supplier: TCI America
Danger

C0110-500ML C0110-25ML
TCC0110-500MLEA 94 USD
TCC0110-500ML TCC0110-25ML
3-Chloroaniline ≥99.0%
3-Chloroaniline

CAS Number: 108-42-9
MDL Number: MFCD00007765
Molecular Formula: C6H6ClN
Molecular Weight: 127.57
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Melting point (°C): -10
Flash Point (°C): 118
Specific Gravity (20/20): 1.22

Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 230 °C
Melting Pt: –10 °C
Density: 1.22 (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00007765
CAS Number: 108-42-9
EINECS: 203-581-0
UN: 2019
ADR: 6.1,II
Merck Index: 13,02136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e62291c23d97e1a37887aefec36759f402acfb9c&vid=062dde078833da276838fbdd94b0db5baa8b2720&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR