Dimethyl carbonate ≥98.0%
Supplier: TCI America
|
Danger
|
CAS Number: 616-38-6
MDL Number: MFCD00008420
Molecular Formula: C3H6O3
Molecular Weight: 90.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Melting point (°C): 4
Flash Point (°C): 17
Specific Gravity (20/20): 1.07
Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification), Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Carbonic Acid Dimethyl Ester
Formula:
C₃H₆O₃ Boiling Pt: 90 °C Melting Pt: 4 °C Density: 1.07 (20 °C) Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008420 CAS Number: 616-38-6 EINECS: 210-478-4 UN: 1161 ADR: 3,II Merck Index: 12,06117 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...