L(-)-Carnitine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
C0049-5G
C0049-25G
TCC0049-005GEA
49.85
USD
TCC0049-005G
TCC0049-025G
L(-)-Carnitine ≥98.0% (by titrimetric analysis)
L(-)-Carnitine
CAS Number: 541-15-1
MDL Number: MFCD00038747
Molecular Formula: C7H16NO3
Molecular Weight: 161.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -32 deg (C=1, H2O)
Product Categories: Bioscience, Vitamin Related Compounds
Other Product Names / Synonyms: (R)-3-Hydroxy-4-(trimethylammonio)butyrate
Formula:
C₇H₁₅NO₃ MW: 161.20 g/mol |
MDL Number:
MFCD00038747 CAS Number: 541-15-1 EINECS: 208-768-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...