Diethyl carbonate ≥98.0%

Supplier: TCI America
Danger

C0041-500G C0041-25G
TCC0041-500GEA 43.27 USD
TCC0041-500G TCC0041-25G
Diethyl carbonate ≥98.0%
Diethyl carbonate

CAS Number: 105-58-8
MDL Number: MFCD00009107
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 25
Specific Gravity (20/20): 0.98


Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification), Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Carbonic Acid Diethyl Ester

Formula: C₅H₁₀O₃
Boiling Pt: 126 °C
Melting Pt: -43 °C
Density: 0.98 (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00009107
CAS Number: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3,III
Merck Index: 13,03815
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=236060f631546feb0243da238b5048a44b96b320&vid=236060f631546feb0243da238b5048a44b96b320&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR