4'-Chloroacetophenone ≥97.0%

Supplier: TCI America
Warning

Synonyms: p-Chloroacetophenone

C0033-25G C0033-100G
TCC0033-25GEA 21.26 USD
TCC0033-25G TCC0033-100G
4'-Chloroacetophenone ≥97.0%
4'-Chloroacetophenone

CAS Number: 99-91-2
MDL Number: MFCD00000624
Molecular Formula: C8H7ClO
Molecular Weight: 154.59
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Flash Point (°C): 118
Freezing point (°C): 15
Specific Gravity (20/20): 1.19

Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 237 °C
Melting Pt: 18…20 °C
Density: 1.19 (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00000624
CAS Number: 99-91-2
EINECS: 202-800-7
UN: 3416
ADR: 6.1,II
Merck Index: 13,02133
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75cd887c3e17f0f1ba1773eec2c6680261439357&vid=75cd887c3e17f0f1ba1773eec2c6680261439357&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR