Carbazole ≥97.0%

Supplier: TCI America
Warning

C0032-25G C0032-100G
TCC0032-25GEA 30.23 USD
TCC0032-25G TCC0032-100G
Carbazole ≥97.0%
Carbazole

CAS Number: 86-74-8
MDL Number: MFCD00004960
Molecular Formula: C12H9N
Molecular Weight: 167.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 355
Melting point (°C): 245
Flash Point (°C): 220

Formula: C₁₂H₉N
MW: 167.21 g/mol
Boiling Pt: 355 °C
Melting Pt: 245 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 220 °C
MDL Number: MFCD00004960
CAS Number: 86-74-8
EINECS: 201-696-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02f2c214e28850f1f72c502772974b93649d827e&vid=02f2c214e28850f1f72c502772974b93649d827e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR