Urethane ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Carbamic acid ethyl ester, Ethyl carbamate
C0028-25G
C0028-500G
C0028-100G
TCC0028-25GEA
16.28
USD
TCC0028-25G
TCC0028-500G
TCC0028-100G
Urethane ≥98.0%
Urethane
CAS Number: 51-79-6
MDL Number: MFCD00007966
Molecular Formula: C3H7NO2
Molecular Weight: 89.09
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Boiling point (°C): 184
Melting point (°C): 50
Flash Point (°C): 92
Other Product Names / Synonyms: Carbamic Acid Ethyl Ester, Ethyl Urethane, Urethane
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Boiling Pt: 182 Melting Pt: 50 °C Density: 1.11 g/cm³ (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00007966 CAS Number: 51-79-6 EINECS: 200-123-1 UN: 2757 ADR: 6.1 II Merck Index: 12,10013 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...