D-(+)-Camphor ≥98.0%

Supplier: TCI America
Warning

Synonyms: (R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, (1R,4R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1R,4R)-(+)-1,7,7-Trimethyl-2-norbornanone, (R)-(+)-1,7,7-Trimethyl-2-norbornanone, (1R)-(+)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (+)-Camphor

C0010-500G C0010-25G
TCC0010-500GEA 173 USD
TCC0010-500G TCC0010-25G
D-(+)-Camphor ≥98.0%
D-(+)-Camphor

CAS Number: 464-49-3
MDL Number: MFCD00064149
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 204
Melting point (°C): 178
Flash Point (°C): 66
Specific rotation [a]20/D: 44.5 deg (C=20, EtOH)

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C
Melting Pt: 178 °C
Density: 0.992 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00064149
CAS Number: 464-49-3
EINECS: 207-355-2
UN: 2717
ADR: 4.1,III
Merck Index: 13,01739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b36cbc8b7ec5a9a93862bffbdd9039f557b87ba2&vid=221f7e46fae534b110730bca3200e402a52ba621&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR