Calcium cyanamide

Supplier: TCI America

C0006-500G C0006-25G
TCC0006-500GEA 47 USD
TCC0006-500G TCC0006-25G
Calcium cyanamide
Calcium cyanamide

CAS Number: 156-62-7
MDL Number: MFCD00064894
Molecular Formula: CCaN2
Molecular Weight: 80.10
Form: Crystal
Color: Gray

Formula: CCaN₂
MDL Number: MFCD00064894
CAS Number: 156-62-7
EINECS: 205-861-8
UN: 1403
ADR: 4.3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8f432aac00270d483292a9b66e56feec53321c&vid=8a8f432aac00270d483292a9b66e56feec53321c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR