Caffeic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

C0002-5G C0002-25G
TCC0002-5GEA 43.9 USD
TCC0002-5G TCC0002-025G
Caffeic acid ≥98.0% (by titrimetric analysis)
Caffeic acid

CAS Number: 331-39-5
MDL Number: MFCD00004392
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Phenylpropanoids, Phenylpropanoids (Phytochemicals), Epigenetics Research Reagents
Other Product Names / Synonyms: 3,4-Dihydroxycinnamic Acid

Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 234…237 °C
MDL Number: MFCD00004392
CAS Number: 331-39-5
EINECS: 206-361-2
Merck Index: 12,01673
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd3f726f6494f95d297c649a33ebd58fe97ebfcb&vid=fd3f726f6494f95d297c649a33ebd58fe97ebfcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR