Benzyl alcohol ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Phenylmethanol
, Benzenemethanol
, Phenylcarbinol
B2378-500G
TCB2378-500GEA
40.31
USD
TCB2378-500G
Benzyl alcohol ≥99.0%
Benzyl alcohol
CAS Number: 100-51-6
MDL Number: MFCD00004599
Molecular Formula: C7H8O
Molecular Weight: 108.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 200
Melting point (°C): -15
Flash Point (°C): 100
Specific Gravity (20/20): 1.05
Formula:
C₆H₅CH₂OH MW: 108.14 g/mol Boiling Pt: 200 °C Melting Pt: –15 °C Density: 1.05 (20 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 Merck Index: 13,01126 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...