2-Bromo-5-fluorophenol ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Bromo-4-fluoro-2-hydroxybenzene
B2040-5G
B2040-25G
TCB2040-005GEA
68
USD
TCB2040-005G
TCB2040-25G
2-Bromo-5-fluorophenol ≥98.0%
2-Bromo-5-fluorophenol
CAS Number: 147460-41-1
MDL Number: MFCD00040939
Molecular Formula: C6H4BrFO
Molecular Weight: 191.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 87
Specific Gravity (20/20): 1.73
Formula:
C₆H₄BrFO Boiling Pt: 184…186 °C (1013 hPa) Density: 1.73 (20 °C) Flash Pt: 87 °C |
MDL Number:
MFCD00040939 CAS Number: 147460-41-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...