N,N'-Bis(3-aminopropyl)ethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,5,8,12-Tetraazadodecane

B1952-500ML B1952-25ML
TCB1952-500MLEA 108.84 USD
TCB1952-500ML TCB1952-25ML
N,N'-Bis(3-aminopropyl)ethylenediamine ≥98.0% (by GC, titration analysis)
N,N'-Bis(3-aminopropyl)ethylenediamine

CAS Number: 10563-26-5
MDL Number: MFCD00008210
Molecular Formula: C8H22N4
Molecular Weight: 174.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 142
Specific Gravity (20/20): 0.95

Formula: C₈H₂₂N₄
MW: 174.29 g/mol
Boiling Pt: 165 °C
Density: 0.95 (20 °C)
Flash Pt: 142 °C
MDL Number: MFCD00008210
CAS Number: 10563-26-5
EINECS: 234-147-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4650cb4293343c75c6a1aa312ee32924382358e&vid=a4650cb4293343c75c6a1aa312ee32924382358e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR