2-(4-Bromophenyl)ethanol ≥97.0%

Supplier: TCI America

Synonyms: 4-Bromophenethyl alcohol

B1918-5G B1918-25G
TCB1918-005GEA 49 USD
TCB1918-005G TCB1918-025G
2-(4-Bromophenyl)ethanol ≥97.0%
2-(4-Bromophenyl)ethanol

CAS Number: 4654-39-1
MDL Number: MFCD00002897
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Specific Gravity (20/20): 1.48


Other Product Names / Synonyms: 4-Bromophenethyl Alcohol, 2-(4-Bromophenyl)ethanol

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 150 °C
Melting Pt: 36 - 38 °C
Density: 1.48 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002897
CAS Number: 4654-39-1
EINECS: 225-093-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec77c92b9d2c042474625c718464f134b027d993&vid=ec77c92b9d2c042474625c718464f134b027d993&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR