3,5-Bis(trifluoromethyl)benzonitrile ≥98.0%

Supplier: TCI America

B1896-25G B1896-5G
TCB1896-25GEA 240 USD
TCB1896-25G TCB1896-005G
3,5-Bis(trifluoromethyl)benzonitrile ≥98.0%
3,5-Bis(trifluoromethyl)benzonitrile

CAS Number: 27126-93-8
MDL Number: MFCD00000379
Molecular Formula: C9H3F6N
Molecular Weight: 239.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 73
Freezing point (°C): 21

Formula: C₉H₃F₆N
MW: 239.12 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.42 g/cm³ (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00000379
CAS Number: 27126-93-8
EINECS: 248-240-7
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9d2cb956267f48ca37e8a21c49db21327e71955&vid=c9d2cb956267f48ca37e8a21c49db21327e71955&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR