4-Bromo-2-fluorophenol ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Bromo-3-fluoro-4-hydroxybenzene
B1835-25G
B1835-5G
TCB1835-025GEA
93
USD
TCB1835-025G
TCB1835-5G
4-Bromo-2-fluorophenol ≥98.0%
4-Bromo-2-fluorophenol
CAS Number: 2105-94-4
MDL Number: MFCD00011722
Molecular Formula: C6H4BrFO
Molecular Weight: 191.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 98
Specific Gravity (20/20): 1.76
Formula:
C₆H₄BrFO Boiling Pt: 90 °C Density: 1.76 (20 °C) Flash Pt: 98 °C |
MDL Number:
MFCD00011722 CAS Number: 2105-94-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...