1-Bromo-4-hexylbenzene ≥90.0%

Supplier: TCI America

Synonyms: (4-Bromophenyl)hexane

B1606-25G B1606-5G
TCB1606-25GEA 178 USD
TCB1606-25G TCB1606-005G
1-Bromo-4-hexylbenzene ≥90.0%
1-Bromo-4-hexylbenzene

CAS Number: 23703-22-2
MDL Number: MFCD00061114
Molecular Formula: C12H17Br
Molecular Weight: 241.17
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Hexylbromobenzene

Formula: C₁₂H₁₇Br
MW: 241.17 g/mol
Boiling Pt: 145 °C
Density: 1.18 (20 °C)
MDL Number: MFCD00061114
CAS Number: 23703-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b2fd9cfe9bd7a8ec204bc831e5c0a673e4cd3b3&vid=5b2fd9cfe9bd7a8ec204bc831e5c0a673e4cd3b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR