4-Biphenylacetonitrile ≥95.0%

Supplier: TCI America

B1545-5G B1545-25G
TCB1545-5GEA 109.8 USD
TCB1545-5G TCB1545-025G
4-Biphenylacetonitrile ≥95.0%
4-Biphenylacetonitrile

CAS Number: 31603-77-7
MDL Number: MFCD00016403
Molecular Formula: C14H11N
Molecular Weight: 193.25
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 93


Other Product Names / Synonyms: 4-Cyanomethyldiphenyl

Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00016403
CAS Number: 31603-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=564abbc444ddffa29a7105751416304037653a70&vid=564abbc444ddffa29a7105751416304037653a70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR