tert-Butyl bromoacetate ≥98.0%

Supplier: TCI America

Synonyms: Bromoacetic tert-butyl ester

B1473-250G B1473-25G
TCB1473-250GEA 324 USD
TCB1473-250G TCB1473-25G
tert-Butyl bromoacetate ≥98.0%
tert-Butyl bromoacetate

CAS Number: 5292-43-3
MDL Number: MFCD00000188
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 49
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: Bromoacetic Acid tert-Butyl Ester

Formula: BrCH₂COOC(CH₃)₂
Boiling Pt: 76 °C
Density: 1.33 (20 °C)
Flash Pt: 49 °C
MDL Number: MFCD00000188
CAS Number: 5292-43-3
EINECS: 226-133-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f4d7c239c54edfaeb33138007fbb73ab7f1ebec&vid=1f4d7c239c54edfaeb33138007fbb73ab7f1ebec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR