Ethyl-6-bromohexanoate ≥98.0%

Supplier: TCI America

B1465-25G B1465-250G
TCB1465-025GEA 54.85 USD
TCB1465-025G TCB1465-250G
Ethyl-6-bromohexanoate ≥98.0%
Ethyl-6-bromohexanoate

CAS Number: 25542-62-5
MDL Number: MFCD00000270
Molecular Formula: C8H15BrO2
Molecular Weight: 223.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 58
Specific Gravity (20/20): 1.26


Other Product Names / Synonyms: 6-Bromohexanoic Acid Ethyl Ester

Formula: BrH₂C(CH₂)₄COOCH₂CH₃
Boiling Pt: 130 °C
Melting Pt: 33 °C
Density: 1.26 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00000270
CAS Number: 25542-62-5
EINECS: 247-085-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a33d18198286344526cabb3d5d1c553ff8c5beac&vid=a33d18198286344526cabb3d5d1c553ff8c5beac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR