Ethyl bromodifluoroacetate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-bromo-2,2-difluoroacetate, Bromodifluoroacetic ethyl ester

B1463-250G B1463-5G
TCB1463-250GEA 311.23 USD
TCB1463-250G TCB1463-5G
Ethyl bromodifluoroacetate ≥98.0%
Ethyl bromodifluoroacetate

CAS Number: 667-27-6
MDL Number: MFCD00042069
Molecular Formula: C4H5BrF2O2
Molecular Weight: 202.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 21
Specific Gravity (20/20): 1.57


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Bromodifluoroacetic Acid Ethyl Ester

Formula: BrF₂CCOOCH₂CH₃
MW: 202.98 g/mol
Boiling Pt: 112 °C (1013 hPa)
Density: 1.57 (20 °C)
Flash Pt: 21 °C
MDL Number: MFCD00042069
CAS Number: 667-27-6
EINECS: 211-567-0
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1abf3b23cf648df1cdfa1455740619ff5814b16d&vid=1abf3b23cf648df1cdfa1455740619ff5814b16d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR