3,6-Dioxaoctamethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Bis-NH2-PEG2, 2,2'-(Ethylenedioxy)bis(ethylamine), 1,8-Diamino-3,6-dioxaoctane, 1,2-Bis(2-aminoethoxy)ethane

B1431-25G B1431-100G
TCB1431-25GEA 47.58 USD
TCB1431-25G TCB1431-100G
3,6-Dioxaoctamethylenediamine ≥98.0% (by GC, titration analysis)
3,6-Dioxaoctamethylenediamine

CAS Number: 929-59-9
MDL Number: MFCD00040474
Molecular Formula: C6H16N2O2
Molecular Weight: 148.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 121
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Monomers, Bioscience, Crosslinkers, PEGylation Reagents & PEG Spacers
Other Product Names / Synonyms: 1,8-Diamino-3,6-dioxaoctane, Ethylene Glycol Bis(2-aminoethyl) Ether

Formula: C₆H₁₆N₂O₂
MW: 148.20 g/mol
Boiling Pt: 123 °C
Density: 1.01 (20 °C)
Flash Pt: 121 °C
MDL Number: MFCD00040474
CAS Number: 929-59-9
EINECS: 213-203-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1bd20f5a67f7c4dcf747154ea6c57083eb7dc62&vid=f1bd20f5a67f7c4dcf747154ea6c57083eb7dc62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR