2-Bromo-2-butene ≥98.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with copper

B1412-500G B1412-25G
TCB1412-500GEA 545 USD
TCB1412-500G TCB1412-25G
2-Bromo-2-butene ≥98.0% stabilized
2-Bromo-2-butene

(stabilized with Copper chip)
CAS Number: 13294-71-8
MDL Number: MFCD00000141
Molecular Formula: C4H7Br
Molecular Weight: 135.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 18
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: 1-Methyl-1-propenyl Bromide

Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 82 °C (1013 hPa)
Density: 1.33 (20 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000141
CAS Number: 13294-71-8
EINECS: 236-316-2
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e02926de0d9da2bf93085f9fc587ddc21404b7e&vid=9589f19bc3b6f1188f18e5a134d85b30afdacb5b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR