α,α,α,α',α',α'-Hexafluoro-p-xylene ≥99.0%

Supplier: TCI America

B1408-500G B1408-25G
TCB1408-500GEA 253 USD
TCB1408-500G TCB1408-25G
α,α,α,α',α',α'-Hexafluoro-p-xylene ≥99.0%
α,α,α,α',α',α'-Hexafluoro-p-xylene

CAS Number: 433-19-2
MDL Number: MFCD00000402
Molecular Formula: C8H4F6
Molecular Weight: 214.11
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 22
Freezing point (°C): -1
Specific Gravity (20/20): 1.39


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-p-xylene

Formula: C₈H₄F₆
Boiling Pt: 117 °C
Melting Pt: 3 °C
Density: 1.39 (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00000402
CAS Number: 433-19-2
EINECS: 207-086-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d83842a8935d7c1e3a6e3023ebe1df816e62b45e&vid=d83842a8935d7c1e3a6e3023ebe1df816e62b45e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR