Benzyltributylammonium chloride ≥98.0%

Supplier: TCI America
Warning

Synonyms: N-Benzyl-N,N-dibutylbutan-1-aminium chloride

B1384-500G B1384-25G
TCB1384-500GEA 99 USD
TCB1384-500G TCB1384-25G
Benzyltributylammonium chloride ≥98.0%
Benzyltributylammonium chloride

CAS Number: 23616-79-7
MDL Number: MFCD00011849
Molecular Formula: C19H34ClN
Molecular Weight: 311.94
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 164

Formula: C₁₉H₃₄ClN
MW: 311.94 g/mol
Melting Pt: 164 °C
MDL Number: MFCD00011849
CAS Number: 23616-79-7
EINECS: 245-787-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0671bc8f13e0d65f6184d21a47c5e9e1ffa3ec6&vid=e0671bc8f13e0d65f6184d21a47c5e9e1ffa3ec6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR