Boc-Pro-OH ≥99.0%

Supplier: TCI America

Synonyms: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid

B1188-5G B1188-25G
TCB1188-005GEA 20 USD
TCB1188-005G TCB1188-025G
Boc-Pro-OH ≥99.0%
Boc-Pro-OH

CAS Number: 15761-39-4
MDL Number: MFCD00037324
Molecular Formula: C10H17NO4
Molecular Weight: 215.25
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 136
Specific rotation [a]20/D: -60 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-proline, Boc-Pro-OH

Formula: C₁₀H₁₇NO₄
Melting Pt: 136 °C
MDL Number: MFCD00037324
CAS Number: 15761-39-4
EINECS: 239-848-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fb8c42ed047037abd2f93a661ea7715a585bd20&vid=5fb8c42ed047037abd2f93a661ea7715a585bd20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR