N-tert-Butylbenzylamine ≥96.0%
Supplier: TCI America
B1176-500ML
B1176-25ML
TCB1176-500MLEA
454
USD
TCB1176-500ML
TCB1176-25ML
N-tert-Butylbenzylamine ≥96.0%
N-tert-Butylbenzylamine
CAS Number: 3378-72-1
MDL Number: MFCD00008798
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 252
Flash Point (°C): 90
Specific Gravity (20/20): 0.90
Formula:
C₁₁H₁₇N Boiling Pt: 252 °C Density: 0.90 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00008798 CAS Number: 3378-72-1 EINECS: 222-179-6 UN: 2735 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...