Bromopentafluorobenzene ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1-Bromo-2,3,4,5,6-pentafluorobenzene

B1116-25G B1116-5G
TCB1116-025GEA 48.1 USD
TCB1116-025G TCB1116-005G
Bromopentafluorobenzene ≥99.0%
Bromopentafluorobenzene

CAS Number: 344-04-7
MDL Number: MFCD00000287
Molecular Formula: C6BrF5
Molecular Weight: 246.96
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Melting point (°C): -31
Flash Point (°C): 87
Specific Gravity (20/20): 1.95


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Pentafluorobromobenzene

Formula: C₆BrF₅
MW: 246.96 g/mol
Boiling Pt: 137 °C
Melting Pt: -31 °C
Density: 1.95 (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000287
CAS Number: 344-04-7
EINECS: 206-449-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41c069f9cbfc783cb954067bcb9532f6c2162ed5&vid=2a8dd0b95a6c05cbefd1252ae373cd81dfea995f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR