Benzopinacol ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: 1,1,2,2-Tetraphenyl-1,2-ethanediol, 1,1,2,2-Tetraphenylethane-1,2-diol, Tetraphenylethylene glycol, Tetraphenyl-1,2-ethanediol

B1037-25G B1037-5G
TCB1037-025GEA 96 USD
TCB1037-025G TCB1037-005G
Benzopinacol ≥95.0% (by HPLC)
Benzopinacol

CAS Number: 464-72-2
MDL Number: MFCD00004448
Molecular Formula: C26H22O2
Molecular Weight: 366.46
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 181


Other Product Names / Synonyms: 1,1,2,2-Tetraphenyl-1,2-ethanediol, Tetraphenylethylene Glycol

Formula: (C₆H₅)₂C(OH)C(OH)(C₆H₅)₂
MW: 366.46 g/mol
Boiling Pt: 508 °C (1013 hPa)
Melting Pt: 181 °C
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00004448
CAS Number: 464-72-2
EINECS: 207-356-8
Merck Index: 13,01101
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b814d242efefd0c3622b6635af89d30e0ba4903f&vid=b814d242efefd0c3622b6635af89d30e0ba4903f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR