1-Benzyl-4-piperidone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone
B1027-25ML
B1027-500ML
TCB1027-025MLEA
31
USD
TCB1027-025ML
TCB1027-500ML
1-Benzyl-4-piperidone ≥98.0% (by GC, titration analysis)
1-Benzyl-4-piperidone
CAS Number: 3612-20-2
MDL Number: MFCD00006192
Molecular Formula: C12H15NO
Molecular Weight: 189.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 71
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C
Formula:
C₁₂H₁₅NO MW: 189.26 g/mol Boiling Pt: 158 °C Density: 1.06 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00006192 CAS Number: 3612-20-2 EINECS: 222-782-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...