4-Butylphenol ≥96.0%

Supplier: TCI America
Danger

B1026-25G B1026-5G
TCB1026-025GEA 177.8 USD
TCB1026-025G TCB1026-005G
4-Butylphenol ≥96.0%
4-Butylphenol

CAS Number: 1638-22-8
MDL Number: MFCD00041750
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 245
Freezing point (°C): 22

Formula: C₁₀H₁₄O
Boiling Pt: 245 °C
Melting Pt: 23…24 °C
Density: 0.9717 g/cm³ (20 °C)
Flash Pt: 121 °C
MDL Number: MFCD00041750
CAS Number: 1638-22-8
EINECS: 216-672-5
UN: 3145
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c3c0836548a5a93be145d2eb9000d9aef4b769b&vid=5bdf90ba2ccb483e1a1e9546d85751adb3b05d2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR