α,4-Dibromotoluene ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromobenzyl bromide, 1-Bromo-4-(bromomethyl)benzene

B1002-25G B1002-100G
TCB1002-25GEA 25 USD
TCB1002-25G TCB1002-100G
α,4-Dibromotoluene ≥98.0%
α,4-Dibromotoluene

CAS Number: 589-15-1
MDL Number: MFCD00000179
Molecular Formula: C7H6Br2
Molecular Weight: 249.93
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 124
Melting point (°C): 61


Other Product Names / Synonyms: alpha,4-Dibromotoluene

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 124 °C
Melting Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00000179
CAS Number: 589-15-1
EINECS: 209-636-5
UN: 2811
ADR: 6.1,II
Merck Index: 13,01393
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c67ec2e3e037e5d0114e18661b170e0be1a808e&vid=76ad614d4eec886e91cbc7ec6d390042cf67bf1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR